You Searched For: Bins
75  results were found
SearchResultCount:"75"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (129-2618)
Supplier: DESCO
Description: This conductive waste bin is useful in large ESD protected areas where waste accumulates and cannot be conveniently removed except in bulk.
UOM: 1 * 1 items
Supplier: Hospidex
Description: Sharpsafe®, historic pioneer in protecting against the risk of sharps injuries and sustainability in medical waste packaging management, presents the ...
Energy Efficient
Low Manufacturing Impact
Product Transparency and Disclosure
Safer Human and Environmental Health
Sustainable Materials
Waste Reducing
Supplier: Palbam
Description: Stainless steel 304, electropolished finish.
Supplier: BOCHEM
Description: Made from stainless steel 18/10, electro-polished.
Supplier: Hospidex
Description: Made from 30% recycled plastic. Fully opening lid, with original positioning of hinge.
Catalog Number: (BOCH11424)
Supplier: BOCHEM
Description: Stainless steel ¹⁸/₁₀ housing.
UOM: 1 * 1 items
Catalog Number: (HUARSHA-41912430)
Supplier: Hospidex
Description: The Clinisafe® plastic clinical waste containers have been designed to hold clinical, anatomical and laboratory waste.
UOM: 1 * 20 items
Supplier: Palbam
Description: This disposal cabinet can be used as a stand-alone unit or in combination with the dispenser unit (connecting frame required).
Supplier: Rubbermaid Commercial Products
Description: A recycling solution with a 'front of house' look and 'back of house' functionality.
Catalog Number: (129-1951)
Supplier: Rubbermaid Commercial Products
Description: Space-efficient, economical, easy and effective way to recycle.
UOM: 1 * 1 items
Supplier: JUSTRITE MANUFACTURING CO
Description: These leakproof sturdy steel containers with foot-operated and self-closing lid may be used to store contaminated laundry as well as other regulated w...
Supplier: Rubbermaid Commercial Products
Description: The Configure™ decorative waste containers provide a recycling solution with sleek, smooth surfaces and contoured edges. This recycling system has a m...
Supplier: Rubbermaid Commercial Products
Description: Space-efficient, economical wastebaskets offer an easy and effective way to recycle.
Sale
Supplier: Rubbermaid Commercial Products
Description: Stationary containers provide convenient central collection sites for multiple work stations or areas.
Supplier: Rubbermaid Commercial Products
Description: Highly durable waste containers created with the toughest enviroments in mind.
Sale
Supplier: Rubbermaid Commercial Products
Description: Safely collect waste wih touch-free closed lid containers.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 75
no targeter for Bottom