You Searched For: Batteries
24  results were found
SearchResultCount:"24"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (611-8684)
Supplier: VWR Collection
Description: Eco friendly battery pack to replace lead-acid batteries which are now restricted/prohibited for transportation.
UOM: 1 * 1 items
Catalog Number: (630-1637)
Supplier: VWR Collection
Description: Solar charger with polymer lithium-ion battery.
UOM: 1 * 1 items
Supplier: VARTA GERAETEBATTERIE
Description: Battery, Lithium batteries, Micro AAA, Voltage: 1,5 V, Ø×H: 10,5×44,5 mm
Supplier: VARTA GERAETEBATTERIE
Description: Batteries, Alkaline battery, LR1, Voltage: 1,5 V
Supplier: VARTA GERAETEBATTERIE
Description: Battery, For: General use, Longlife alkaline manganese battery, 9 V-Block/6LR61
Catalog Number: (111-2548)
Supplier: 3M
Description: The fast-charging, intrinsically safe Versafloâ„¢ lithium-ion battery TR 830 is intended for use with the 3Mâ„¢ Versafloâ„¢ TR 800 series PAPR system.
UOM: 1 * 1 items
Supplier: VARTA GERAETEBATTERIE
Description: Long life batteries for cameras and clocks.
Supplier: VARTA GERAETEBATTERIE
Description: Long life batteries. For use in electronic appliances, calculators, cameras, and more.
Supplier: VARTA GERAETEBATTERIE
Description: High capacity with long operating times per charge. Do not require a special charger.
Catalog Number: (111-2369)
Supplier: 3M
Description: Intelligent charger that cuts off the charging process when battery is fully charged. For use with the Jupiterâ„¢ powered air respirator battery pack.
UOM: 1 * 1 items
Supplier: Testo
Description: Batteries, USB connection cable for 175/176
Catalog Number: (MMMATR-644E)
Supplier: 3M
Description: The Versafloâ„¢ Battery Charger Kit TR-644E is a four-station battery charger designed for use with all batteries from the Versafloâ„¢ Powered Air Turbo T...
UOM: 1 * 1 items
Catalog Number: (139-2271)
Supplier: VARTA GERAETEBATTERIE
Description: Nickel Hydride (NiMH), AA/HR6 1,2 V. These batteries are compatible with a wide range of cordless phones including Alcatel, Ascom, Audioline, Binatone...
UOM: 1 * 2 items
Catalog Number: (111-2549)
Supplier: 3M
Description: The Versafloâ„¢ Battery Charger Kit TR-641E is a single-station battery charger which consists of a TR-640 charging cradle, power supply, and lead. It's...
UOM: 1 * 1 items
Catalog Number: (MMMATR-632)
Supplier: 3M
Description: The Versafloâ„¢ High Capacity Battery TR-632 is a rechargeable lithium-ion battery designed for use with the Versafloâ„¢ Powered Air Turbo TR-600 Series.
UOM: 1 * 1 items
Supplier: DWK Life Sciences
Description: Batteries, Battery backup, field installed for R2P
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 24
no targeter for Bottom