You Searched For: Waste Disposal Bags
68  results were found
SearchResultCount:"68"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: These sturdy bags are ideal for disposing of biologically hazardous waste that must first be autoclaved. PP, 50 µm thick.
Supplier: VWR Collection
Description: Ideal for processing biohazardous materials that require autoclaving prior to disposal. Available with or without temperature indicator. High integrit...
Supplier: VWR Collection
Description: Sturdy bags, ideal for disposing of biologically hazardous waste that must first be autoclaved. Complete with wire closures that allow expansion of th...
Supplier: VWR Collection
Description: Sturdy, temperature resistant bags, ideal for disposing of hazardous waste that must first be autoclaved. Can also be used for autoclaving other produ...
Supplier: VWR Collection
Description: Sturdy bags, ideal for disposing of non hazardous waste that must first be autoclaved. In PP, 50 µm thick, transparent, with labelling area.
Supplier: VWR Collection
Description: Sturdy bags, ideal for disposing of biologically hazardous waste that must first be autoclaved. PP, 75 µm thick, orange, with 'Biohazard' label.
Supplier: VWR Collection
Description: These bags are ideal for autoclaving polystyrene containers and other items contaminated by pathogenic agents (petri dishes, microtitre plates, pipett...
Supplier: VWR Collection
Description: For safe disposal of laboratory waste such as pipette tips, disposable Petri dishes, sample tubes, etc. PP, 40 µm thick, clear, with or without 'Bioha...
Supplier: VWR Collection
Description: Constructed of durable PP with feature the universal biohazard symbol in black.
Sale
Supplier: VWR Collection
Description: Designed for safe and secure transport of a specimen container and documentation, these 51 µm thick bags feature a pouch to keep the paperwork separat...
Supplier: VWR Collection
Description: These bags are ideal for autoclaving polystyrene containers and other items contaminated by pathogenic agents (petri dishes, microtitre plates, pipett...
Supplier: VWR Collection
Description: Bucket liners create an impermeable barrier between the disinfectant and receptacle.
Supplier: VWR Collection
Description: Sturdy bags, ideal for disposing of biologically hazardous waste that must first be autoclaved. Supplied complete with the necessary sealing wires. HD...
Catalog Number: (129-1575)
Supplier: VWR Collection
Description: Sturdy, temperature resistant bags, ideal for sterilising animal cages.
UOM: 1 * 250 items
Supplier: KEYSTONE CLEANROOM PRODUCTS
Description: Ready to use pre-sterilised trash bags make it easy to remove waste from pharmaceutical production areas without introducing potential sources of cont...
Supplier: KEYSTONE CLEANROOM PRODUCTS
Description: Reduce the number of janitorial steam sterilisation loads by using gamma irradiated mop bucket liners. Instead of before every cleaning, the mop bucke...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 68
no targeter for Bottom