You Searched For: Aluminium Foils
12  results were found
SearchResultCount:"12"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Pure aluminium 99,0 to 99,5%.
Supplier: ALUJET
Description: Pure aluminium 99,0 to 99,5%. Ideal for packaging, covering and insulation.
Supplier: Korff
Description: Pure aluminium, 99,0 to 99,5%.
Supplier: Korff
Description: Pure aluminium, 99,0 to 99,5%, soft.
Supplier: Korff
Description: Pure aluminium 99,0 to 99,5%, soft.
Catalog Number: (KORF70237)
Supplier: Korff
Description: This aluminum foil is packed on rolls of about 5 kg.
UOM: 1 * 1 Roll
Supplier: Korff
Description: Aluminium foil, square cut.
Catalog Number: (293-4321)
Supplier: Korff
Description: Pure aluminium 99,0 to 99,5%.
UOM: 1 * 200 items
Catalog Number: (GERB5712)
Supplier: GERBER FUNKE DR N.
Description: These aluminium foils are used with reference drier RD-8 for determining the moisture content of powdered milk.
UOM: 1 * 1.000 items
Catalog Number: (293-4109)
Supplier: MELITTA HAUSHALTPRODUKTE
Description: Ideal for packaging, covering and insulation. Can be used for a wide variety of laboratory tasks.
UOM: 1 * 1 Roll
Supplier: VWR Products
Description: Designed for use in controlled environments, ultra high vacuum aluminium foil is virtually free from contamination and residual rolling oils. Foil is ...
Catalog Number: (MELL4008871200839)
Supplier: MELITTA HAUSHALTPRODUKTE
Description: Toppits® Cling Film is ideal for keeping food fresh with the unique bow-shaped tear-off edge and the ergonomic packaging allows particularly easy hand...
UOM: 1 * 1 Roll
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 12 of 12
no targeter for Bottom