You Searched For: Sprays
7  results were found
SearchResultCount:"7"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (BOHLH958-04)
Supplier: Bohlender
Description: Release and anti-blocking agent as well as lubricant for laboratory. Excellent gliding and separating effects.
UOM: 1 * 1 items
MSDS
Catalog Number: (291-1246)
Supplier: HM INDUSTRIESERVICE GMBH
Description: Cleaner spray for electronic parts, 200 ml
UOM: 1 * 200 mL
Catalog Number: (291-1204)
Supplier: HM INDUSTRIESERVICE GMBH
Description: Label remover for hassle-free removal of paper labels.
UOM: 1 * 200 mL
Supplier: HM INDUSTRIESERVICE GMBH
Description: For the quick and easy removal of dust and small amounts of dirt in hard to reach places. For use in medical engineering, metrology, microscopy, preci...
Catalog Number: (KINGLE511964)
Supplier: KING DISPOSABLES
Description: Suma Rapid D6 is a quick drying glass cleaner for use on windows, mirrors, display cabinets, tiles and other glazed ceramics.
UOM: 1 * 1 items
Catalog Number: (291-1270)
Supplier: DRAWIN VERTRIEBS GMBH
Description: Silicone spray AK is an multi-purpose release agent and lubricant for use in industry and laboratory.
UOM: 1 * 200 mL
Catalog Number: (291-1240)
Supplier: HM INDUSTRIESERVICE GMBH
Description: Prevents and eliminates electrostatic charges and their interference.
UOM: 1 * 200 mL
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 7 of 7
no targeter for Bottom