You Searched For: Adapters, Spherical Ground Joints
14  results were found
SearchResultCount:"14"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Lenz Laborglas GmbH & CO.KG
Description: DURAN®, borosilicate glass, clear.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Made from DURAN® tubing.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Made of stainless steel.
Supplier: Lenz Laborglas GmbH & CO.KG
Description: Made of DURAN®, borosilicate glass.
Supplier: VACUUBRAND GMBH + CO
Description: These small flanges with female ground joint are made of stainless steel.
Supplier: VACUUBRAND GMBH + CO
Description: These small flanges made of stainless steel.
Supplier: DWK Life Sciences
Description: DURAN®, borosilicate glass 3.3, clear.
Supplier: DWK Life Sciences
Description: DURAN®, borosilicate glass 3.3, clear.
Supplier: DWK Life Sciences
Description: DURAN®, borosilicate glass 3.3, clear.
Supplier: DWK Life Sciences
Description: DURAN®, borosilicate glass 3.3, clear.
Catalog Number: (WITG0.998.129)
Supplier: WITEG LABORTECHNIK GMBH
Description: Borosilicate glass.
UOM: 1 * 1 items
Supplier: SciLabware
Description: Made of PYREX®, borosilicate glass.
Supplier: SciLabware
Description: Made of borosilicate glass.
Sale
Supplier: JUCHHEIM Laborgerate GmbH
Description: For reaction vessels, 316 stainless steel, thread length 14 mm.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 14 of 14
no targeter for Bottom