Order Entry
Multidishes, poly-D-lysine coated, Nunc™
undefined
Multidishes, poly-D-lysine coated, Nunc™
  • Catalog No:
  • 735-0227
  • NUNC152025
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  •  
    Seq
    No. of wells
    Colour
    Culture area
    Recommended working volume
    Sterile
  • Multidishes, PDL-coated, aseptic
    Multidishes, PDL-coated, aseptic
    NUNC152025
    152025
    01
    24
    Clear
    1,9 cm²
    1 ml/well
    -
    1 * 20 items
    1 * 20 items - €574.00
    €574.00
    1 * 20 items
       €574.00
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Multidishes, PDL-coated
    Multidishes, PDL-coated
    735-0227
    152035
    02
    6
    Clear
    9,6 cm²
    3 ml/well
    -
    1 * 20 items
    1 * 20 items - €660.00
    €660.00
    1 * 20 items
       €660.00
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact