Nitroterephthalic acid 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-Nitroterephthalic acid
CAS Number:610-29-7
:
Formula:C₈H₅NO₆
MDL Number:MFCD00007141
Molecular Weight:211.13 g/mol
Melting Pt:270…272 °C
EINECS:210-217-4
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to light beige Powder
- Infrared spectrum:Conforms
- Titration with NaOH:≥99.0 %
Specification Test Results