Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (4)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Hand Cleanser and Soap Dispensers
SearchPresentationType-VERTICAL
SearchResultCount:"25"
  List View Grid View Easy View (new)
Sort by:
Catalog Number: (129-0486)
Supplier:  VWR Collection
Description:   The Touch-Free soap dispenser reduces cross-contamination and waste in cleanrooms and supporting areas.
UOM:  1 * 1 items
Supplier:  SCA
Description:   These liquid hand soap dispensers are designed for kitchen and shower areas.
Supplier:  SCA
Description:   These dispensers with their modern design are suitable for a high image washroom interior.
Supplier:  Deb-STOKO
Description:   This dispenser made from steel is available with or without long arm lever.
Catalog Number: (115-0648)
Supplier:  Deb-STOKO
Description:   Rugged plastic housing make this dispenser easy to use and to refill in showers.
UOM:  1 * 1 items
Catalog Number: (ZVGA60810-11)
Supplier:  Zellstoff-Vertriebs-GmbH & Co. KG
Description:   This durable zetMatic® dispenser is made from robust plastic. It can be perfectly integrated into every washroom.
UOM:  1 * 1 items
Catalog Number: (ZVGA60814-02)
Supplier:  Zellstoff-Vertriebs-GmbH & Co. KG
Description:   The long life soap dispenser can be filled with all off-the-shelf liquid soaps.
UOM:  1 * 1 items
Catalog Number: (115-1513)
Supplier:  Deb-STOKO
Description:   Hand cleanser dispenser, Aluminium, Grey/white, 1000 ml, 94×162×316 mm
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 25
  1  2  3  4  Next