Order Entry
Optical Glass Filters
Optical Glass Filters
Certificates
undefined
Optical Glass Filters
Related Information
Filters for focusing lens
Product Information
  • Catalog #
    Supplier #
    Seq
    For
    Pk
    Availability
    Price
  • Daylight filter, Ø 17 mm
    Daylight filter, Ø 17 mm
    631-0010
    158211
    02
    Focusing lens (631-0009, 631-0007) and KL1500HAL cold light source
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 46,00 €
    46,00 €
    New Customer Promotional Price
    1 * 1 items
       46,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Halogen filter, Ø 17 mm
    Halogen filter, Ø 17 mm
    630-1459
    158207
    01
    Focusing lens (631-0009, 631-0007) and KL300 or KL1600LED or KL2500LED cold light source
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 46,00 €
    46,00 €
    New Customer Promotional Price
    1 * 1 items
       46,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
Filter holder
Product Information
  • Catalog #
    Supplier #
    Seq
    For
    Pk
    Availability
    Price
  • Filter holder (empty), for insert filters Ø 28 mm
    Filter holder (empty), for insert filters Ø 28 mm
    SCOC258330
    258330
    KL1600LED/2500LED/KL1500HAL
    5
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items - 60,00 €
    60,00 €
    New Customer Promotional Price
    1 * 5 items
       60,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
Insert filters
Product Information
  • Catalog #
    Supplier #
    Seq
    For
    Pk
    Availability
    Price
  • Colour filter green, Ø 28 mm
    Colour filter green, Ø 28 mm
    631-0525
    258304
    06
    KL1600LED/2500LED/KL1500HAL
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 41,00 €
    41,00 €
    New Customer Promotional Price
    1 * 1 items
       41,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Colour filter blue, Ø 28 mm
    Colour filter blue, Ø 28 mm
    631-0523
    258302
    04
    KL1600LED/2500LED/KL1500HAL
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 62,00 €
    62,00 €
    New Customer Promotional Price
    1 * 1 items
       62,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Colour filter red, Ø 28 mm
    Colour filter red, Ø 28 mm
    631-0524
    258303
    05
    KL1600LED/2500LED/KL1500HAL
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 41,00 €
    41,00 €
    New Customer Promotional Price
    1 * 1 items
       41,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Colour filter yellow, Ø 28 mm
    Colour filter yellow, Ø 28 mm
    631-0526
    258305
    07
    KL1600LED/2500LED/KL1500HAL
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 41,00 €
    41,00 €
    New Customer Promotional Price
    1 * 1 items
       41,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Daylight filter, Ø 28 mm
    Daylight filter, Ø 28 mm
    SCOC258306
    258306
    09
    KL1500HAL
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 41,00 €
    41,00 €
    New Customer Promotional Price
    1 * 1 items
       41,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Blue fluorescense excitation filter, Ø 28 mm
    Blue fluorescense excitation filter, Ø 28 mm
    SCOC258313
    258313
    11
    KL1600LED/2500LED/KL1500HAL
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 414,00 €
    414,00 €
    New Customer Promotional Price
    1 * 1 items
       414,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Halogen filter, Ø 28 mm
    Halogen filter, Ø 28 mm
    631-0916
    258307
    10
    KL1600LED/2500LED
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 41,00 €
    41,00 €
    New Customer Promotional Price
    1 * 1 items
       41,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
Insert filters
Product Information
  • Catalog #
    Seq
    Description
    For
    Pk
    Availability
    Price
  • 04
    Colour filter blue, Ø 28 mm
    KL1600LED/2500LED/KL1500HAL
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 62,00 €
    62,00 €
    New Customer Promotional Price
Product Information
  • Catalog #
    Seq
    Description
    For
    Pk
    Availability
    Price
  • 07
    Colour filter yellow, Ø 28 mm
    KL1600LED/2500LED/KL1500HAL
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 41,00 €
    41,00 €
    New Customer Promotional Price
Product Information
  • Catalog #
    Seq
    Description
    For
    Pk
    Availability
    Price
  • 10
    Halogen filter, Ø 28 mm
    KL1600LED/2500LED
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 41,00 €
    41,00 €
    New Customer Promotional Price
Product Information
  • Catalog #
    Seq
    Description
    For
    Pk
    Availability
    Price
  • 09
    Daylight filter, Ø 28 mm
    KL1500HAL
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 41,00 €
    41,00 €
    New Customer Promotional Price
Product Information
  • Catalog #
    Seq
    Description
    For
    Pk
    Availability
    Price
  • 11
    Blue fluorescense excitation filter, Ø 28 mm
    KL1600LED/2500LED/KL1500HAL
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 414,00 €
    414,00 €
    New Customer Promotional Price
Product Information
  • Catalog #
    Seq
    Description
    For
    Pk
    Availability
    Price
  • 06
    Colour filter green, Ø 28 mm
    KL1600LED/2500LED/KL1500HAL
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 41,00 €
    41,00 €
    New Customer Promotional Price
Product Information
  • Catalog #
    Seq
    Description
    For
    Pk
    Availability
    Price
  • 05
    Colour filter red, Ø 28 mm
    KL1600LED/2500LED/KL1500HAL
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 41,00 €
    41,00 €
    New Customer Promotional Price
Filters for focusing lens
Product Information
  • Catalog #
    Seq
    Description
    For
    Pk
    Availability
    Price
  • 02
    Daylight filter, Ø 17 mm
    Focusing lens (631-0009, 631-0007) and KL1500HAL cold light source
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 46,00 €
    46,00 €
    New Customer Promotional Price
Product Information
  • Catalog #
    Seq
    Description
    For
    Pk
    Availability
    Price
  • 01
    Halogen filter, Ø 17 mm
    Focusing lens (631-0009, 631-0007) and KL300 or KL1600LED or KL2500LED cold light source
    1
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 46,00 €
    46,00 €
    New Customer Promotional Price