3-(3-Aminophenyl)propionic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
L19329.03EA
60.1
EUR
L19329.03
3-(3-Aminophenyl)propionic acid ≥97%
3-(3-Aminophenyl)propionic acid
Formula:
C₉H₁₁NO₂ MW: 165,19 g/mol Melting Pt: 99…101 °C Storage Temperature: Ambient |
EINECS:
000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...