You Searched For: Dehydrated Media for Microbiology
109  results were found
SearchResultCount:"109"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (J869-500G)
Supplier: VWR Chemicals
Description: Terrific broth is an enriched medium, ideal for culturing cells for nucleic acid purification and recombinant protein expression.
UOM: 1 * 500 g
MSDS
Supplier: VWR Chemicals
Description: A range of culture media in compliance with ISO standards and the International Harmonised Pharmacopoeia. All our media are, as you would expect, prod...
Supplier: VWR Chemicals
Description: Dehydrated culture media in small sachets ready to make up exactly 500 ml of medium.
Catalog Number: (J873-500G)
Supplier: VWR Chemicals
Description: Used for the preparation of media for the detection of yeasts and moulds.
UOM: 1 * 500 g
Supplier: VWR Chemicals
Description: VWR® dehydrated culture media that has been granulated offers both convenience and safety benefits. The correct dissolution of culture media is critic...
Catalog Number: (J906-500G)
Supplier: VWR Chemicals
Description: Super optimal broth (SOB) broth is used for the preparation of nutritionally-rich growth medium and transformation of competent E. coli.
UOM: 1 * 500 g
MSDS
Supplier: VWR Chemicals
Description: Scholten's broth (modified)
MSDS Certificates New Product
Supplier: VWR Chemicals
Description: Lysogeny broth (LB) is a nutritionally rich medium used for the propagation and maintenance of E. coli. It is also widely used for the preparat...
Supplier: VWR Chemicals
Description: 2XYT medium broth powder is used to prepare rich broth for the propagation of M13 bacteriophage.
Catalog Number: (3508.)
Supplier: Avantor
Description: Faucet diluting fluid
UOM: 1 * 1 items
MSDS
Catalog Number: (J903-500G)
Supplier: VWR Chemicals
Description: YPD broth is used for the propagation and maintenance of yeasts.
UOM: 1 * 500 g
Supplier: THERMO FISHER DIAGNOSTICS GMBH
Description: Thioglycolate broth, 500 g, Application: Differential media, Selective for: Anaerobs, Standards: USP
Supplier: MP Biomedicals
Description: Rich medium for the growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium.
Supplier: Thermo Scientific
Description: Terrific Broth
Supplier: Thermo Scientific
Description: YP Base Medium
Supplier: LIOFILCHEM S.R.L.
Description: Dehydrated culture media in powder available in 100 g, 500 g and 5 kg formats.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 109
no targeter for Bottom