Order Entry
SMD Tweezer
New Product
SMD Tweezer
New Product
  •  
    Seq
    Material
    Length
    Tip style
    Tip width
    Thickness
  • SMD tweezers, sandblast coated, height 1,5 mm, net.weight 13,5 g
    SMD tweezers, sandblast coated, height 1,5 mm, net.weight 13,5 g
    OUTISMD08
    SMD08
    01
    Stainless steel
    120 mm
    Curved/angled/bent tips
    2,5 mm
    0,35 mm (tip)
    1 * 1 items
    1 * 1 items - 33,90 €
    33,90 €
    1 * 1 items
       33,90 €
    Specifications
    Environmentally Preferable
  • SMD15 tweezers, sandblast coated, height 1,5 mm, net.weight 13,5 g
    SMD15 tweezers, sandblast coated, height 1,5 mm, net.weight 13,5 g
    OUTISMD15
    SMD15
    02
    Stainless steel
    120 mm
    Curved/angled/bent tips
    2,5 mm
    0,35 mm (tip)
    1 * 1 items
    1 * 1 items - 37,30 €
    37,30 €
    1 * 1 items
       37,30 €
    Specifications
    Environmentally Preferable
  • SMD tweezers, sandblast coated, with serrated handle, height 2 mm, net.weight 27 g
    SMD tweezers, sandblast coated, with serrated handle, height 2 mm, net.weight 27 g
    OUTI572-SA
    572-SA
    03
    Stainless steel
    152 mm
    Straight/arc shaped/grooved
    6,4 mm
    0,4 mm (tip)
    1 * 1 items
    1 * 1 items - 28,50 €
    28,50 €
    1 * 1 items
       28,50 €