Order Entry
Solaris 2000 I Small Incubated Benchtop Orbital Shakers
Solaris 2000 I Small Incubated Benchtop Orbital Shakers
Certificates
undefined
Solaris 2000 I Small Incubated Benchtop Orbital Shakers
Limited Time Sale
Select items below are currently on sale
Specifications
  • Catalog No:
  • 171-1517
  • 444-1436
  • BARNSK2001MPKG
  • Model:
  • Solaris 2000 I
  • Solaris 2000 I
  • Solaris 2000 I
  • Shaking motion:
  • Orbital
  • Orbital
  • Orbital
  • Temperature range:
  • 30…60 °C
  • 30…60 °C
  • 30…60 °C
  • Speed range:
  • 15 - 525 min⁻¹
  • 15 - 525 min⁻¹
  • 15 - 525 min⁻¹
  • Max. load:
  • 16 kg
  • 16 kg
  • 16 kg
  • Warranty:
  • Standard warranty includes 2 years labour, 5 years parts, and 10 years on the drive mechanism
  • Standard warranty includes 2 years labour, 5 years parts, and 10 years on the drive mechanism
  • Standard warranty includes 2 years labour, 5 years parts, and 10 years on the drive mechanism
  • Orbit:
  • 25 mm
  • 25 mm
  • 25 mm
  • Electrical:
  • 100 - 240 V, 50/60 Hz
  • 100 - 240 V, 50/60 Hz
  • 100 - 240 V, 50/60 Hz
  • Display:
  • Touch screen
  • Touch screen
  • Touch screen
Specifications
Product Information
  • Catalog #
    Supplier #
    Seq
    W×D×H
    Pk
    Availability
    Price
  • Solaris 2000 I with 305×356 mm (12×14") platform
    Solaris 2000 I with 305×356 mm (12×14") platform
    444-1436
    SK2001
    01
    701×360×460 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items - 9 240,00 €
    9 240,00 €
    Sale
    Valid until 30/06/2025
    SaleValid until 30/06/2025
    New Customer Promotional Price
    1 * 1 items
       9 240,00 €
    Specifications
    Model
    Solaris 2000 I
    Shaking motion
    Orbital
    Environmentally Preferable
    Temperature range
    30…60 °C
    Speed range
    15 - 525 min⁻¹
    Max. load
    16 kg
    Warranty
    Standard warranty includes 2 years labour, 5 years parts, and 10 years on the drive mechanism
    Orbit
    25 mm
    Electrical
    100 - 240 V, 50/60 Hz
    Display
    Touch screen
  • Solaris 2000 I with 305×356 mm (12×14") platform and flask clamp starter kit
    Solaris 2000 I with 305×356 mm (12×14") platform and flask clamp starter kit
    171-1517
    SK2001CPKG
    02
    701×360×460 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items - 9 800,00 €
    9 800,00 €
    New Customer Promotional Price
    1 * 1 items
       9 800,00 €
    Specifications
    Model
    Solaris 2000 I
    Shaking motion
    Orbital
    Environmentally Preferable
    Temperature range
    30…60 °C
    Speed range
    15 - 525 min⁻¹
    Max. load
    16 kg
    Warranty
    Standard warranty includes 2 years labour, 5 years parts, and 10 years on the drive mechanism
    Orbit
    25 mm
    Electrical
    100 - 240 V, 50/60 Hz
    Display
    Touch screen
  • Solaris 2000 I with 305×356 mm (12×14") platform and adhesive sticky mats
    Solaris 2000 I with 305×356 mm (12×14") platform and adhesive sticky mats
    BARNSK2001MPKG
    SK2001MPKG
    03
    701×360×460 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items - 9 300,00 €
    9 300,00 €
    New Customer Promotional Price
    1 * 1 items
       9 300,00 €
    Specifications
    Model
    Solaris 2000 I
    Shaking motion
    Orbital
    Environmentally Preferable
    Temperature range
    30…60 °C
    Speed range
    15 - 525 min⁻¹
    Max. load
    16 kg
    Warranty
    Standard warranty includes 2 years labour, 5 years parts, and 10 years on the drive mechanism
    Orbit
    25 mm
    Electrical
    100 - 240 V, 50/60 Hz
    Display
    Touch screen