Multi-purpose medium-duty gloves, ActivArmr® 97-008
: Ansell
- Catalog No:
- ANSE97-008/11
- ANSE97-008/9
- ANSE97-008/8
- ANSE97-008/10
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex
- Latex
- Latex
- Latex
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Liner material:
- Nylon/Spandex/Kevlar®
- Nylon/Spandex/Kevlar®
- Nylon/Spandex/Kevlar®
- Nylon/Spandex/Kevlar®
- Finish:
- Palm coated (foam nitrile)
- Palm coated (foam nitrile)
- Palm coated (foam nitrile)
- Palm coated (foam nitrile)
- Gauge:
- 15
- 15
- 15
- 15
- Cuff style:
- Hook & loop/Velcro® fastening
- Hook & loop/Velcro® fastening
- Hook & loop/Velcro® fastening
- Hook & loop/Velcro® fastening
- EN norm:
- EN 388:2016 (4232B)
- EN 388:2016 (4232B)
- EN 388:2016 (4232B)
- EN 388:2016 (4232B)
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- SeqGlove sizeColourLengthPk
- ANSE97-008/897-008/8018Black/grey/orange243 - 284 mm (10 - 11")12 PAIR1 * 12 PAIR434,00 €
Specifications
Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLatex / latex-freeLatexPowdered / powder-freePowder-freeLiner materialNylon/Spandex/Kevlar®FinishPalm coated (foam nitrile)Gauge15Cuff styleHook & loop/Velcro® fasteningEN normEN 388:2016 (4232B)SterilityNon sterile - ANSE97-008/997-008/9029Black/grey/orange243 - 284 mm (10 - 11")12 PAIR1 * 12 PAIR434,00 €
Specifications
Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLatex / latex-freeLatexPowdered / powder-freePowder-freeLiner materialNylon/Spandex/Kevlar®FinishPalm coated (foam nitrile)Gauge15Cuff styleHook & loop/Velcro® fasteningEN normEN 388:2016 (4232B)SterilityNon sterile - ANSE97-008/1097-008/100310Black/grey/orange243 - 284 mm (10 - 11")12 PAIR1 * 12 PAIR434,00 €
Specifications
Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLatex / latex-freeLatexPowdered / powder-freePowder-freeLiner materialNylon/Spandex/Kevlar®FinishPalm coated (foam nitrile)Gauge15Cuff styleHook & loop/Velcro® fasteningEN normEN 388:2016 (4232B)SterilityNon sterile - ANSE97-008/1197-008/110411Black/grey/orange243 - 284 mm (10 - 11")12 PAIR1 * 12 PAIR434,00 €
Specifications
Ambidextrous / hand-specificHand-specificEnvironmentally PreferableDisposable / reusableReusableLatex / latex-freeLatexPowdered / powder-freePowder-freeLiner materialNylon/Spandex/Kevlar®FinishPalm coated (foam nitrile)Gauge15Cuff styleHook & loop/Velcro® fasteningEN normEN 388:2016 (4232B)SterilityNon sterile