Article combination AC 4003, for thermal process control in food and pharma market, EBRO®
Supplier: Ebro
ebro®
620-2834EA 3396 EUR
620-2834
Article combination AC 4003, for thermal process control in food and pharma market, EBRO®
Data Loggers
This combination of 3 data loggers for temperature measurement as well as evaluation software and interface are suitable for process monitoring in a range from −30 to +140 °C. In addition to the measurement data acquisition, the Winlog.pro software offers a variety of analysis options and calculations.
  • To measure the temperature inside the product and control the pasteurisation or sterilisation
The software will show the temperature profile and provides a lot of options to evaluate the measurement data.
Certifikace: EG 1935/2004, 2014/53/EU, 2011/65/EU.
Delivery information: The data loggers includes a 3 point temperature calibration certificate.