Domiphen bromide
Supplier: Spectrum Chemical
Certificates
About this item
Synonyms: (Dodecyldimethyl-2-phenoxyethyl)ammonium bromide, N,N-Dimethyl-N-(2-phenoxyethyl)dodecan-1-aminium bromide
CAS Number:538-71-6
:
Formula:C₂₂H₄₀BrNO
MDL Number:MFCD00011769
Molecular Weight:414.48 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications