Order Entry
Glass crimp top headspace vials, level 3 high performance applications, SureSTART™
Glass crimp top headspace vials, level 3 high performance applications, SureSTART™
Certificates
undefined
Glass crimp top headspace vials, level 3 high performance applications, SureSTART™
Related Information
Vials
Product Information
  • Catalog #
    Supplier #
    Seq
    Capacity
    Base
    Ø×H
    Material
    Colour
    Availability
    Price
  • Crimp vial, headspace
    Crimp vial, headspace
    HYPE6PCV6-1FF
    6PCV6-1FF
    01
    6,00 ml
    Square edge flat bottom
    22×38 mm
    Glass
    Clear
    Log in for availability
    Log in for availability
    1 * 5.000 items
    1 * 5.000 items - 2 260,00 €
    2 260,00 €
    New Customer Promotional Price
    1 * 5.000 items
       2 260,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Crimp vial, headspace
    Crimp vial, headspace
    HYPE6PCV6-1F
    6PCV6-1F
    02
    6,00 ml
    Square edge flat bottom
    22×38 mm
    Glass
    Clear
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 50,60 €
    50,60 €
    New Customer Promotional Price
    1 * 100 items
       50,60 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Crimp vial, headspace
    Crimp vial, headspace
    HYPE6PCV6-1FT
    6PCV6-1FT
    03
    6,00 ml
    Square edge flat bottom
    22×38 mm
    Glass
    Clear
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 486,00 €
    486,00 €
    New Customer Promotional Price
    1 * 1.000 items
       486,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Crimp vial, headspace
    Crimp vial, headspace
    HYPE6PCV10-V1
    6PCV10-V1
    04
    9,50 ml
    Beveled edge flat bottom
    22,75×45,2 mm
    Glass
    Clear
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 458,00 €
    458,00 €
    New Customer Promotional Price
    1 * 100 items
       458,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Crimp vial, headspace
    Crimp vial, headspace
    HYPE6PCV10-1F
    6PCV10-1F
    06
    10,00 ml
    Beveled edge flat bottom
    22,75×46 mm
    Glass
    Clear
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 55,30 €
    55,30 €
    New Customer Promotional Price
    1 * 100 items
       55,30 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Crimp vial, headspace
    Crimp vial, headspace
    HYPE6PCV10-1FT
    6PCV10-1FT
    07
    10,00 ml
    Beveled edge flat bottom
    22,75×46 mm
    Glass
    Clear
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 525,00 €
    525,00 €
    New Customer Promotional Price
    1 * 1.000 items
       525,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Crimp vial, headspace
    Crimp vial, headspace
    HYPE6PCV20-1FF
    6PCV20-1FF
    08
    20,00 ml
    Beveled edge flat bottom
    23×75 mm
    Glass
    Clear
    Log in for availability
    Log in for availability
    1 * 5.000 items
    1 * 5.000 items - 2 570,00 €
    2 570,00 €
    New Customer Promotional Price
    1 * 5.000 items
       2 570,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Crimp vial, headspace
    Crimp vial, headspace
    HYPE6PCV20-1F
    6PCV20-1F
    09
    20,00 ml
    Beveled edge flat bottom
    22,75×75 mm
    Glass
    Clear
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 56,80 €
    56,80 €
    New Customer Promotional Price
    1 * 100 items
       56,80 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Crimp vial, headspace
    Crimp vial, headspace
    HYPE6PCV20-1FT
    6PCV20-1FT
    10
    20,00 ml
    Beveled edge flat bottom
    23×75 mm
    Glass
    Clear
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 534,00 €
    534,00 €
    New Customer Promotional Price
    1 * 1.000 items
       534,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Crimp vial, headspace
    Crimp vial, headspace
    HYPE6PCV20-1RF
    6PCV20-1RF
    11
    20,00 ml
    Beveled edge round bottom
    23×75 mm
    Glass
    Clear
    Log in for availability
    Log in for availability
    1 * 5.000 items
    1 * 5.000 items - 2 700,00 €
    2 700,00 €
    New Customer Promotional Price
    1 * 5.000 items
       2 700,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Crimp vial, headspace
    Crimp vial, headspace
    HYPE6PCV20-1R
    6PCV20-1R
    12
    20,00 ml
    Beveled edge round bottom
    22,75×75 mm
    Glass
    Clear
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 59,90 €
    59,90 €
    New Customer Promotional Price
    1 * 100 items
       59,90 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Crimp vial, headspace
    Crimp vial, headspace
    HYPE6PCV20-1RT
    6PCV20-1RT
    13
    20,00 ml
    Beveled edge round bottom
    23×75 mm
    Glass
    Clear
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 563,00 €
    563,00 €
    New Customer Promotional Price
    1 * 1.000 items
       563,00 €
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
Caps
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 9
    Magnetic crimp cap, with thinned penetration area for SPME
    45 shore
    White silicone/blue PTFE
    3,0 mm
    Blue
    20 mm
    Aluminium/tinplate
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 1 550,00 €
    1 550,00 €
    New Customer Promotional Price
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 10
    Non-magnetic crimp cap
    45 shore
    Blue silicone/clear PTFE, soft
    3,0 mm
    Red
    20 mm
    Aluminium
    Log in for availability
    Log in for availability
    1 * 5.000 items
    1 * 5.000 items - 5 230,00 €
    5 230,00 €
    New Customer Promotional Price
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 12
    Non-magnetic crimp cap
    45 shore
    Blue silicone/clear PTFE, soft
    3,0 mm
    Red
    20 mm
    Aluminium
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 1 100,00 €
    1 100,00 €
    New Customer Promotional Price
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 11
    Non-magnetic crimp cap
    45 shore
    Blue silicone/clear PTFE, soft
    3,0 mm
    Red
    20 mm
    Aluminium
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 116,00 €
    116,00 €
    New Customer Promotional Price
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 1
    Magnetic crimp cap
    45 shore
    Blue silicone/clear PTFE, soft
    3,0 mm
    Silver
    20 mm
    Aluminium/tinplate
    Log in for availability
    Log in for availability
    1 * 5.000 items
    1 * 5.000 items - 3 690,00 €
    3 690,00 €
    New Customer Promotional Price
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 2
    Magnetic crimp cap
    45 shore
    Blue silicone/clear PTFE, soft
    3,0 mm
    Silver
    20 mm
    Aluminium/tinplate
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 81,60 €
    81,60 €
    New Customer Promotional Price
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 4
    Magnetic crimp cap
    54 shore
    Red silicone/natural PTFE, high temperature
    3,0/1,5 mm
    Silver
    20 mm
    Aluminium/tinplate
    Log in for availability
    Log in for availability
    1 * 5.000 items
    1 * 5.000 items - 6 360,00 €
    6 360,00 €
    New Customer Promotional Price
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 5
    Magnetic crimp cap
    45 shore
    Red silicone/natural PTFE, high temperature
    3,0 mm
    Silver
    20 mm
    Aluminium/tinplate
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 145,00 €
    145,00 €
    New Customer Promotional Price
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 3
    Magnetic crimp cap
    45 shore
    Blue silicone/clear PTFE, soft
    3,0 mm
    Silver
    20 mm
    Aluminium/tinplate
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 775,00 €
    775,00 €
    New Customer Promotional Price
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 7
    Magnetic crimp cap, with thinned penetration area for SPME
    54 shore
    White silicone/blue PTFE
    3,0/1,5 mm
    Blue
    20 mm
    Aluminium/tinplate
    Log in for availability
    Log in for availability
    1 * 5.000 items
    1 * 5.000 items - 7 440,00 €
    7 440,00 €
    New Customer Promotional Price
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 6
    Magnetic crimp cap
    45 shore
    Red silicone/natural PTFE, high temperature
    3,0 mm
    Silver
    20 mm
    Aluminium/tinplate
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 1 380,00 €
    1 380,00 €
    New Customer Promotional Price
Product Information
  • Catalog #
    Seq
    Description
    Hardness
    Septum
    Thickness
    Cap colour
    Cap Size
    Material
    Availability
    Price
  • 8
    Magnetic crimp cap, with thinned penetration area for SPME
    45 shore
    White silicone/blue PTFE
    3,0/1,5 mm
    Blue
    20 mm
    Aluminium/tinplate
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 164,00 €
    164,00 €
    New Customer Promotional Price