Order Entry
LEVIFLOW® Single-Use Clamp-On Ultrasonic Flow Sensors
undefined
LEVIFLOW® Single-Use Clamp-On Ultrasonic Flow Sensors
  • Accuracy:
  • ±3%
  • ±3%
  • Max. pressure:
  • 94 psi
  • 94 psi
  • Min. operating temperature:
  • 10 °C (50 °F)
  • 10 °C (50 °F)
  • Max. operating temperature:
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • Viscosity:
  • 0,7 to 10 cSt
  • 0,7 to 10 cSt
  • Wetted materials:
  • None
  • None
  • Warranty:
  • 1 year
  • 1 year
  •  
    Fluid type
    Maximum Flow Rate
    Process connection
    Pk
  • Clamp-On Ultrasonic Flow Sensor; 20 L/min
    Clamp-On Ultrasonic Flow Sensor; 20 L/min
    Liquids flowing through C-Flex® (374, 072, or 082) and silicone (Pharma 50, 65, or 80) tubing
    20 L/min
    12 mm OD tubing
    1
    1 * 1 items
    1 * 1 items - 3 160,00 €
    3 160,00 €
    1 * 1 items
       3 160,00 €
    CE Compliant
    Accessories
    Specifications
    Environmentally Preferable
    Accuracy
    ±3%
    Max. pressure
    94 psi
    Min. operating temperature
    10 °C (50 °F)
    Max. operating temperature
    60 °C (140 °F)
    Viscosity
    0,7 to 10 cSt
    Wetted materials
    None
    Warranty
    1 year
    CE Compliant
  • Clamp-On Ultrasonic Flow Sensor; 4 L/min
    Clamp-On Ultrasonic Flow Sensor; 4 L/min
    Liquids flowing through C-Flex® (374, 072, or 082) and silicone (Pharma 50, 65, or 80) tubing
    4 L/min
    8 mm OD tubing
    1
    1 * 1 items
    1 * 1 items - 3 160,00 €
    3 160,00 €
    1 * 1 items
       3 160,00 €
    CE Compliant
    Accessories
    Specifications
    Environmentally Preferable
    Accuracy
    ±3%
    Max. pressure
    94 psi
    Min. operating temperature
    10 °C (50 °F)
    Max. operating temperature
    60 °C (140 °F)
    Viscosity
    0,7 to 10 cSt
    Wetted materials
    None
    Warranty
    1 year
    CE Compliant