Order Entry
Masterflex® Microbore Transfer Tubing, PTFE, Avantor®
Masterflex® Microbore Transfer Tubing, PTFE, Avantor®
Certificates
undefined
Masterflex® Microbore Transfer Tubing, PTFE, Avantor®
Certificates
Specifications
  • Model:
  • 06417-41
  • 06417-31
  • 06417-51
  • 06417-21
  • 06417-11
  • Clarity:
  • Translucent
  • Translucent
  • Translucent
  • Translucent
  • Translucent
  • GMP compliant:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Length per pack:
  • 30,5 m (100 ft.)
  • 30,5 m (100 ft.)
  • 30,5 m (100 ft.)
  • 30,5 m (100 ft.)
  • 30,5 m (100 ft.)
  • Max. pressure:
  • 275,8 bar (4000 psi)
  • 275,8 bar (4000 psi)
  • 275,8 bar (4000 psi)
  • 275,8 bar (4000 psi)
  • 275,8 bar (4000 psi)
  • Max. temperature:
  • 260 °C (500 °F)
  • 260 °C (500 °F)
  • 260 °C (500 °F)
  • 260 °C (500 °F)
  • 260 °C (500 °F)
  • Min. temperature:
  • –240 °C (–400 °F)
  • –240 °C (–400 °F)
  • –240 °C (–400 °F)
  • –240 °C (–400 °F)
  • –240 °C (–400 °F)
  • Regulatory approvals:
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • Sterilisation method autoclave:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Sterilisation method EtO:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Sterilisation method gamma:
  • No
  • No
  • No
  • No
  • No
  • Working pressure:
  • 275,8 bar (4000 psi)
  • 275,8 bar (4000 psi)
  • 275,8 bar (4000 psi)
  • 275,8 bar (4000 psi)
  • 275,8 bar (4000 psi)
Specifications
Related Information
Product Information
  • Catalog #
    Seq
    Tubing int.Ø
    Wall thickness
    Tubing ext.Ø
    Tubing material
    Length
    Colour
    Availability
    Price
  • Transfer Tubing, Microbore PTFE, 0,012" ID x 0,030" OD; 100 ft/roll
    Transfer Tubing, Microbore PTFE, 0,012" ID x 0,030" OD; 100 ft/roll
    01
    0,3048 mm (0,012")
    0,2286 mm
    0,762 mm (0,03")
    PTFE
    30,5 m
    Translucent
    Log in for availability
    Log in for availability
    1 * 100 Feet
    1 * 100 Feet - 34,20 €
    34,20 €
    1 * 100 Feet
       34,20 €
    Specifications:
    CE certificate
    More
    Specifications
    Model
    06417-11
    Environmentally Preferable
    Clarity
    Translucent
    GMP compliant
    Yes
    Length per pack
    30,5 m (100 ft.)
    Max. pressure
    275,8 bar (4000 psi)
    Max. temperature
    260 °C (500 °F)
    Min. temperature
    –240 °C (–400 °F)
    Regulatory approvals
    FDA;USP
    Sterilisation method autoclave
    Yes
    Sterilisation method EtO
    Yes
    Sterilisation method gamma
    No
    Working pressure
    275,8 bar (4000 psi)
    CE certificate
  • Transfer Tubing, Microbore PTFE, 0,022" ID x 0,042" OD; 100 ft/roll
    Transfer Tubing, Microbore PTFE, 0,022" ID x 0,042" OD; 100 ft/roll
    02
    0,5588 mm (0,022")
    0,254 mm
    1,0668 mm (0,042")
    PTFE
    30,5 m
    Translucent
    Log in for availability
    Log in for availability
    1 * 100 Feet
    1 * 100 Feet - 37,60 €
    37,60 €
    1 * 100 Feet
       37,60 €
    Specifications:
    CE certificate
    More
    Specifications
    Model
    06417-21
    Environmentally Preferable
    Clarity
    Translucent
    GMP compliant
    Yes
    Length per pack
    30,5 m (100 ft.)
    Max. pressure
    275,8 bar (4000 psi)
    Max. temperature
    260 °C (500 °F)
    Min. temperature
    –240 °C (–400 °F)
    Regulatory approvals
    FDA;USP
    Sterilisation method autoclave
    Yes
    Sterilisation method EtO
    Yes
    Sterilisation method gamma
    No
    Working pressure
    275,8 bar (4000 psi)
    CE certificate
  • Transfer Tubing, Microbore PTFE, 0,032" ID x 0,056" OD; 100 ft/roll
    Transfer Tubing, Microbore PTFE, 0,032" ID x 0,056" OD; 100 ft/roll
    03
    0,8128 mm (0,032")
    0,3048 mm
    1,4224 mm (0,056")
    PTFE
    30,5 m
    Translucent
    Log in for availability
    Log in for availability
    1 * 100 Feet
    1 * 100 Feet - 54,60 €
    54,60 €
    1 * 100 Feet
       54,60 €
    Specifications:
    CE certificate
    More
    Specifications
    Model
    06417-31
    Environmentally Preferable
    Clarity
    Translucent
    GMP compliant
    Yes
    Length per pack
    30,5 m (100 ft.)
    Max. pressure
    275,8 bar (4000 psi)
    Max. temperature
    260 °C (500 °F)
    Min. temperature
    –240 °C (–400 °F)
    Regulatory approvals
    FDA;USP
    Sterilisation method autoclave
    Yes
    Sterilisation method EtO
    Yes
    Sterilisation method gamma
    No
    Working pressure
    275,8 bar (4000 psi)
    CE certificate
  • Transfer Tubing, Microbore PTFE, 0,042" ID x 0,066" OD; 100 ft/roll
    Transfer Tubing, Microbore PTFE, 0,042" ID x 0,066" OD; 100 ft/roll
    04
    1,0668 mm (0,042")
    0,3048 mm
    1,6764 mm (0,066")
    PTFE
    30,5 m
    Translucent
    Log in for availability
    Log in for availability
    1 * 100 Feet
    1 * 100 Feet - 48,30 €
    48,30 €
    1 * 100 Feet
       48,30 €
    Specifications:
    CE certificate
    More
    Specifications
    Model
    06417-41
    Environmentally Preferable
    Clarity
    Translucent
    GMP compliant
    Yes
    Length per pack
    30,5 m (100 ft.)
    Max. pressure
    275,8 bar (4000 psi)
    Max. temperature
    260 °C (500 °F)
    Min. temperature
    –240 °C (–400 °F)
    Regulatory approvals
    FDA;USP
    Sterilisation method autoclave
    Yes
    Sterilisation method EtO
    Yes
    Sterilisation method gamma
    No
    Working pressure
    275,8 bar (4000 psi)
    CE certificate
  • Transfer Tubing, Microbore PTFE, 0,052" ID x 0,076" OD; 100 ft/roll
    Transfer Tubing, Microbore PTFE, 0,052" ID x 0,076" OD; 100 ft/roll
    05
    1,3208 mm (0,052")
    0,3048 mm
    1,9304 mm (0,076")
    PTFE
    30,5 m
    Translucent
    Log in for availability
    Log in for availability
    1 * 100 Feet
    1 * 100 Feet - 69,60 €
    69,60 €
    1 * 100 Feet
       69,60 €
    Specifications:
    CE certificate
    More
    Specifications
    Model
    06417-51
    Environmentally Preferable
    Clarity
    Translucent
    GMP compliant
    Yes
    Length per pack
    30,5 m (100 ft.)
    Max. pressure
    275,8 bar (4000 psi)
    Max. temperature
    260 °C (500 °F)
    Min. temperature
    –240 °C (–400 °F)
    Regulatory approvals
    FDA;USP
    Sterilisation method autoclave
    Yes
    Sterilisation method EtO
    Yes
    Sterilisation method gamma
    No
    Working pressure
    275,8 bar (4000 psi)
    CE certificate