Order Entry
Masterflex® Luer Fittings on PVC Flexible Tubing, Straight, Avantor®
Masterflex® Luer Fittings on PVC Flexible Tubing, Straight, Avantor®
undefined
Masterflex® Luer Fittings on PVC Flexible Tubing, Straight, Avantor®
  • Model:
  • 30600-67
  • 30600-62
  • 30600-60
  • 30526-18
  • 30526-14
  • 30526-16
  • Connection type (port 1):
  • Male luer lock
  • Male luer lock
  • Female luer
  • Female luer
  • Female luer
  • Female luer
  • Connection type (port 2):
  • Male luer lock
  • Male luer lock
  • Male luer lock
  • Male luer lock
  • Male luer lock
  • Male luer lock
  • No. of ports:
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • Warranty:
  • Out of box
  • Out of box
  • Out of box
  • Out of box
  • Out of box
  • Out of box
  •  
    Seq
    Connection Type
    Fitting material
    Flow pattern
    Pk
  • Fitting, Polycarbonate, Straight, Flexible PVC Tubing with Female/Male Luer Ends, 10" Long; 10/PK
    Fitting, Polycarbonate, Straight, Flexible PVC Tubing with Female/Male Luer Ends, 10" Long; 10/PK
    01
    Tubing Assembly
    Polycarbonate
    Straight
    10
    1 * 10 items
    1 * 10 items - 31,40 €
    31,40 €
    1 * 10 items
       31,40 €
    Specifications
    Model
    30526-14
    Environmentally Preferable
    Connection type (port 1)
    Female luer
    Connection type (port 2)
    Male luer lock
    No. of ports
    2
    Warranty
    Out of box
  • Fitting, Polycarbonate, Straight, Flexible PVC Tubing with Female/Male Luer Ends, 20" Long; 10/PK
    Fitting, Polycarbonate, Straight, Flexible PVC Tubing with Female/Male Luer Ends, 20" Long; 10/PK
    02
    Tubing Assembly
    Polycarbonate
    Straight
    10
    1 * 10 items
    1 * 10 items - 30,10 €
    30,10 €
    1 * 10 items
       30,10 €
    Specifications
    Model
    30526-16
    Environmentally Preferable
    Connection type (port 1)
    Female luer
    Connection type (port 2)
    Male luer lock
    No. of ports
    2
    Warranty
    Out of box
  • Fitting, Polycarbonate, Straight, Flexible PVC Tubing with Female/Male Luer Ends, 48" Long; 10/PK
    Fitting, Polycarbonate, Straight, Flexible PVC Tubing with Female/Male Luer Ends, 48" Long; 10/PK
    03
    Tubing Assembly
    Polycarbonate
    Straight
    10
    1 * 10 items
    1 * 10 items - 57,70 €
    57,70 €
    1 * 10 items
       57,70 €
    Specifications
    Model
    30526-18
    Environmentally Preferable
    Connection type (port 1)
    Female luer
    Connection type (port 2)
    Male luer lock
    No. of ports
    2
    Warranty
    Out of box
  • Fitting, Polycarbonate, Straight, Flexible PVC Tubing with Female/Male Luer Ends, 24" Long; 10/PK
    Fitting, Polycarbonate, Straight, Flexible PVC Tubing with Female/Male Luer Ends, 24" Long; 10/PK
    04
    Tubing Assembly
    Polycarbonate
    Straight
    10
    1 * 10 items
    1 * 10 items - 32,70 €
    32,70 €
    1 * 10 items
       32,70 €
    Specifications
    Model
    30600-60
    Environmentally Preferable
    Connection type (port 1)
    Female luer
    Connection type (port 2)
    Male luer lock
    No. of ports
    2
    Warranty
    Out of box
  • Fitting, Polycarbonate, Straight, Flexible PVC Tubing with Male/Male Luer Ends, 24" Long; 10/PK
    Fitting, Polycarbonate, Straight, Flexible PVC Tubing with Male/Male Luer Ends, 24" Long; 10/PK
    06
    Tubing Assembly
    Polycarbonate
    Straight
    10
    1 * 10 items
    1 * 10 items - 34,10 €
    34,10 €
    1 * 10 items
       34,10 €
    Specifications
    Model
    30600-62
    Environmentally Preferable
    Connection type (port 1)
    Male luer lock
    Connection type (port 2)
    Male luer lock
    No. of ports
    2
    Warranty
    Out of box
  • PVC tubing with luer ends, 24" L, M/M fitting ends; 20/pack, Non-sterile
    PVC tubing with luer ends, 24" L, M/M fitting ends; 20/pack, Non-sterile
    08
    Tubing Assembly
    PVC
    Straight
    20
    1 * 20 items
    1 * 20 items - 72,70 €
    72,70 €
    1 * 20 items
       72,70 €
    Specifications
    Model
    30600-67
    Environmentally Preferable
    Connection type (port 1)
    Male luer lock
    Connection type (port 2)
    Male luer lock
    No. of ports
    2
    Warranty
    Out of box