NUCLEIC-CARD™ Nucleic Acid Stabilisation and Archiving Devices
Supplier: Copan
Certificates
About this item
Specifications
- Application:
- Method / Format:
- Refine target molecule:
- Sample type:
- Target molecule:
- Environmentally Preferable:
Specifications
Nucleic Card
- SeqPk
- NUCLEIC-CARD™ 1 spot, 300×6, Packet/BatchNUCLEIC-CARD™ 1 spot, 300×6, Packet/BatchCOPA3001C503001C5021.8001 * 1.800 items6.740,00 €
- NUCLEIC-CARD™ 1 spot, 100×36, Packet/BatchNUCLEIC-CARD™ 1 spot, 100×36, Packet/BatchCOPA3001C1003001C1003.6001 * 3.600 items13.370,00 €
- NUCLEIC-CARD™ 1 spot, framed, 50×36, boxedNUCLEIC-CARD™ 1 spot, framed, 50×36, boxedCOPA40U032D40U032D41 * 1.800 items8.660,00 €
- NUCLEIC-CARD™ 2 spots, 100×20, Packet/BatchNUCLEIC-CARD™ 2 spots, 100×20, Packet/BatchCOPA3005C1003005C10051 * 2.000 items7.930,00 €
- NUCLEIC-CARD™ 4 spots, 100×100, Packet/BatchNUCLEIC-CARD™ 4 spots, 100×100, Packet/BatchCOPA3006C1003006C10061.0001 * 1.000 items5.200,00 €
Sample Collection Device (kit)
- SeqPk
- NUCLEIC-CARD™ sample collection device, 50×1 boxedNUCLEIC-CARD™ sample collection device, 50×1 boxedCOPA40U031D40U031D7501 * 50 items370,00 €