You Searched For: Power Supplies
21  results were found
SearchResultCount:"21"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (700-0856)
Supplier: VWR Collection
Description: VWR® mini power supply is a basic, economical unit perfect for small nucleic acid gel tanks.
UOM: 1 * 1 items
Catalog Number: (MFLX77200-07)
Supplier: Avantor Fluid Handling
Description: Make the most of your Masterflex drives replacement parts and other accessories.
UOM: 1 * 1 items
Catalog Number: (MFLX77120-11)
Supplier: Avantor Fluid Handling
Description: Safely adapt your VDC powered C/L® pump.
UOM: 1 * 1 items
CE Compliant
Catalog Number: (700-1316)
Supplier: VWR Collection
Description: VWR® PerfectBlue power supply is constructed with flame-retardant ABC face plate and is ideal for DNA, RNA, and protein electrophoresis, as well as bl...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: Power supply, VWR®, US-plug, Power supply, dual voltage
Catalog Number: (700-1285)
Supplier: VWR Collection
Description: VWR® PerfectBlue power supply is constructed with PC housing and flame-retardant ABS face plate, and is ideal for DNA, RNA and protein electrophoresis...
UOM: 1 * 1 items
Supplier: VWR Collection
Description: For most electrophoresis applications, the Basic power supply is ideal. For high current blotting applications, choose the High Current power supply.
Catalog Number: (MFLX07525-90)
Supplier: Avantor Fluid Handling
Description: Optimize performance of your Masterflex pump.
UOM: 1 * 1 items
Supplier: Corning
Description: Axygen power supplies are ideal for most electrophoresis needs.
Catalog Number: (115-3060)
Supplier: DESCO
Description: PCB rack is moulded from a conductive thermoplastic compound based on polypropylene copolymer with carbon black and reinforced with mica flake.
UOM: 1 * 1 items
Catalog Number: (NANOPAC-300P)
Supplier: NANOENTEK
Description: The nanoPAC mini power supply are an ultra compact and economic unit ideal for use with DNA (horizontal) and protein (vertical) electrophoresis system...
UOM: 1 * 1 items
New Product
Supplier: SCHOTT AG Lighting and imaging
Description: Power supply, Power cord EU, 1,8 m
Catalog Number: (VART57976101111)
Supplier: VARTA GERAETEBATTERIE
Description: This is a powerful power bank with a capacity of 10000 mAh.
UOM: 1 * 1 items
Supplier: Cytiva
Description: EPS power supplies cover a wide range of electrophoresis and blotting applications. Designed for performance, EPS power supplies can be relied on for ...
Catalog Number: (PSA410)
Supplier: Cytiva
Description: Connects 4 mm output jack to 4 mm high voltage leads.
UOM: 1 * 1 KIT
Supplier: Cytiva
Description: Choice of adapters for connecting 4 or 2 mm leads to 2 or 4 mm output jacks, respectively.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 21
no targeter for Bottom