You Searched For: Tissue Baths
5  results were found
SearchResultCount:"5"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (720-2423)
Supplier: VWR Collection
Description: The reliable W20 histology water bath is ideal for removing wrinkles and distortions in paraffin wax sections which are created during sectioning. The...
UOM: 1 * 1 items
Catalog Number: (462-0624)
Supplier: LAUDA
Description: Tissue float bath for stretching and drying cut tissue samples. Working temperature range of the histology bath is 25 to 80 °C.
UOM: 1 * 1 items
Catalog Number: (ELET48572-55)
Supplier: Cole-Parmer
Description: Designed to assist in tissue preparation in histology and pathology laboratories.
UOM: 1 * 1 items
Catalog Number: (BOEI145951-2)
Supplier: BOEKEL SCIENTIFIC
Description: Precision temperature controlled water bath that eliminates wrinkles and distortion during the preparation of paraffin embedded tissue sections.
UOM: 1 * 1 items
Supplier: Slee
Description: This tissue float bath is designed for stretching of paraffin sections in histology, research and industry laboratories.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 5 of 5
no targeter for Bottom