Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Fluorescence Microscopes
https://si.vwr.com/stibo/smallweb/std.lang.all/52/01/18945201.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"8"
  List View Easy View (new)
Sort by:
Supplier:  VWR Collection
Description:   LED microscope for transmitted and fluorescence reflected light.
Supplier:  VWR Collection
Description:   The VisiScope IT405 FLD has been designed for tissue culture research in clinical and biotechnology laboratories.
Supplier:  VWR Collection
Description:   This fluorescence microscope is designed for lab applications.
UOM:  1 * 1 items
Supplier:  DISCOVER ECHO
Description:   This hybrid microscope easily converts between upright/inverted and brightfield/fluorescence configurations.
Supplier:  ZEISS
Description:   The Axio Vert A1 FL-LED has a four-position mount for LED modules for reflected light fluorescence. One position is occupied for green fluorescent pro...
Supplier:  Molecular Devices
Description:   The ImageXpress Pico system combines powerful analysis for individual labs that need easy automated imaging solutions. Intelligent cell imaging and an...
Catalog Number: (630-2749)
Supplier:  IDCP B.V.
Description:   This handheld microscope is designed for research and observation of fluorescent objects by using 575 nm excitation LEDs.
UOM:  1 * 1 items
Catalog Number: (IDCPAM4515T4-GFBW)
Supplier:  IDCP B.V.
Description:   This handheld microscope is designed for research and observation of fluorescent objects.
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 8
A new look and a renewed commitment
Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.
Learn More Start Shopping