You Searched For: Syringeless Filters
7  results were found
SearchResultCount:"7"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Sartorius
Description: Claristep® filter units with PP housing and RC membranes are used for sample preparation by particle removal prior to analytics. The filters are suita...
Supplier: Whatman products (Cytiva)
Description: Mini-UniPrepâ„¢ G2 is an 'all in one' syringeless filter with a borosilicate glass chamber. It replaces syringe filters for autosampler-supported UHPLC/...
Supplier: Restek
Description: SINGLE StEP® eXtreme filter vials are recommended for samples containing less than 30% solid particulates and also offers fast sample filtration with ...
Catalog Number: (REST28309)
Supplier: Restek
Description: Thomson SINGLE StEP low evaporation filter vials with screw-top caps, 0,45 µm, PTFE with low-evaporation cap, blue cap, 100 pack.
UOM: 1 * 100 items
New Product
Supplier: Restek
Description: Standard filter vials are recommended for samples containing less than 10% solid particulates and also offers fast sample filtration with a squeeze of...
Supplier: Whatman products (Cytiva)
Description: UniPrepâ„¢ Syringeless Filters are preassembled filtration devices for the filtration and storage of laboratory samples. These devices are quick and eas...
Supplier: Whatman products (Cytiva)
Description: The Whatman Mini-UniPrep Syringeless Filters combine four products into one syringeless filter solution for efficient chromatography sample preparatio...
Sale
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 7 of 7
no targeter for Bottom