You Searched For: Fall Harnesses
25  results were found
SearchResultCount:"25"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: 3M
Description: 3M DBI-SALA ExoFit XE200 harness with rescue loops, automatic buckles, two sizes. X-shaped chest straps with central D-ring attachment point optimised...
New Product
Supplier: 3M
Description: Cross chest buckle assembly for central front fall arrest attachment point - optimised position on body. Revolving torso adjusters (patented) for quic...
New Product
Supplier: CAPITAL SAFETY DEUTSCHLAND
Description: The Protecta® Comfort Belt Style Fall Arrest Harness is designed to provide reliable protection, comfort, and convenience. It now features upgraded fe...
Supplier: CAPITAL SAFETY DEUTSCHLAND
Description: Protecta® Standard Vest Style Fall Arrest Harness is a sleek and modern safety harness with upgraded features. It has water and stain resistant webbin...
Catalog Number: (PRTWFP10RER)
Supplier: Portwest
Description: Fall Protection is put in place to prevent the risks associated with falling from heights, reducing impact force, restricting obstacle/ground collisio...
UOM: 1 * 1 items
Supplier: CAPITAL SAFETY DEUTSCHLAND
Description: Protecta® fall arrest harness accessory options provide the opportunity to add extra tool carrying convenience and comfort to a harness.
Catalog Number: (PRTWFP14RER)
Supplier: Portwest
Description: Fall Protection is put in place to prevent the risks associated with falling from heights, reducing impact force, restricting obstacle/ground collisio...
UOM: 1 * 1 items
Catalog Number: (CPSDAM450/80)
Supplier: CAPITAL SAFETY DEUTSCHLAND
Description: The Protecta® Nylon Webbing Sling Anchor features four safety seams, and measures 80 cm in length.
UOM: 1 * 5 items
Catalog Number: (PRTWFP13RER)
Supplier: Portwest
Description: Fall Protection is put in place to prevent the risks associated with falling from heights, reducing impact force, restricting obstacle/ground collisio...
UOM: 1 * 1 items
Catalog Number: (PRTWFP11RER)
Supplier: Portwest
Description: Fall Protection is put in place to prevent the risks associated with falling from heights, reducing impact force, restricting obstacle/ground collisio...
UOM: 1 * 1 items
Catalog Number: (PRTWFP18RER)
Supplier: Portwest
Description: Fall Protection is put in place to prevent the risks associated with falling from heights, reducing impact force, restricting obstacle/ground collisio...
UOM: 1 * 1 items
Catalog Number: (PRTWFP12RER)
Supplier: Portwest
Description: Fall Protection is put in place to prevent the risks associated with falling from heights, reducing impact force, restricting obstacle/ground collisio...
UOM: 1 * 1 items
Catalog Number: (113-1274)
Supplier: HONEYWELL SAFETY
Description: This harness is ideal for industrial rope access, rescue and special forces. It can be used in conjunction with a climber's seat and an ascender.
UOM: 1 * 1 items
Catalog Number: (113-1250)
Supplier: HONEYWELL SAFETY
Description: This detachable climbers seat offers a comfortable supported position for the operator during prolonged periods suspended at height. It is made from m...
UOM: 1 * 1 items
Supplier: HONEYWELL SAFETY
Description: Miller H-Design® Duraflex™ 2 points harness with two webbing loops at the front and automatic buckles on chest and leg straps. The Miller H-Design® ha...
Supplier: HONEYWELL SAFETY
Description: This harness is the perfect solution for work at height in explosive atmospheres. It is particularly suitable for petrochemical and mining environment...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 25
no targeter for Bottom