Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (14)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
underwear
https://si.vwr.com/stibo/smallweb/std.lang.all/99/66/18989966.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"33"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Collection
Description:   The Quantus® underwear trousers are made from a high-quality breathable polyester microfibre material with carbon. The fabric was specially developed ...
Supplier:  VWR Collection
Description:   The Quantus® underwear T-shirt is made from a breathable polyester microfibre material with carbon. The fabric was specially developed for use as unde...
Catalog Number: (113-8817)
Supplier:  VWR Collection
Description:   This disposable inner wear made from non woven comfortable SMS consists of a shirt and a pair of trousers. The set can be used as innerwear suits or w...
UOM:  1 * 100 SET
Supplier:  VWR Collection
Description:   The Quantus® underwear sweatshirt is made from a high-quality breathable polyester microfibre material with carbon. The fabric was specially developed...
Supplier:  SIOEN PERSONAL PROTECTIVE CLOT
Description:   T-shirts made of SIO-FIT® Thermal wear: 50% Viloft® and 50% polyester (interlock). The fabric keeps the body warm and dry. Ideal for use as a basic la...
Supplier:  SIOEN PERSONAL PROTECTIVE CLOT
Description:   Anti-static and flame-retardant underwear made of soft handle knitted fabric consisting of 60% Protex, 39% Viloft and 1% AST (Jersey).
Supplier:  SIOEN PERSONAL PROTECTIVE CLOT
Description:   These long johns and T-shirts are made of breathable PP fabric. They offer thermal insulation, breathability and moisture management properties.
Supplier:  SIOEN PERSONAL PROTECTIVE CLOT
Description:   These long johns are made of soft SIO-COOL® Comfort polyester, an anti-microbial fabric eliminating all odours. The underwear features moisture absorb...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 33
  1  2  3  4  5  Next