You Searched For: Detergents and Surface Cleansers
167  results were found
SearchResultCount:"167"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: Compact HDPE canisters (Cat. Nos. 845xx.365) are compatible with Miele dosing cabinets.
Catalog Number: (00699030.)
Supplier: VWR Chemicals
Description: Acidic neutraliser and pre-cleaner based on organic acid.
UOM: 1 * 5 kg
Supplier: VWR Chemicals
Description: A range of cleaning agents designed for use in ultrasonic baths.
MSDS Sale
Catalog Number: (7348.5000)
Supplier: Avantor
Description: RBS 35 concentrate is used for manual cleaning, soaking and ultrasonic baths. These surfactants are alkaline, foaming and insensitive to water hardnes...
UOM: 1 * 5 L
Supplier: VWR Chemicals
Description: Iso-propyl alcohol Ph. Eur. 70% v/v, with purified water Ph. Eur. 30% v/v, filtered at 0,2 μm.
Catalog Number: (94006.1016)
Supplier: VWR Chemicals
Description: Iso-propyl alcohol Ph. Eur. 70% V/V, with purified water Ph. Eur. 30% V/V, filtered at 0,2 μm.
UOM: 1 * 6 L
Catalog Number: (3451.1500)
Supplier: Avantor
Description: Cleaning application: Wax repellent, Paraffin cleaner
UOM: 1 * 1.500 mL
MSDS
Catalog Number: (88289.360)
Supplier: VWR Chemicals
Description: Alkaline, powerful universal cleaning agent for use in ultrasonic baths or for manual cleaning in an immersion bath.
UOM: 1 * 5 kg
MSDS
Catalog Number: (2157-04)
Supplier: MACRON (A AVANTOR BRAND)
Description: CHEM-SOLV® is a non chromic acid cleaner suitable for removing silicone greases, dried blood, and other organic residues from laboratory glass and pla...
UOM: 1 * 1 items
Supplier: Diversey
Description: ClearKlens Plus contains a concentrated blend of detergents and emulsifiers. Its balanced neutral formulation is particularly effective against produc...
Catalog Number: (DIVY7513050)
Supplier: Diversey
Description: Ammonia-free, glass cleaner to give a non streaking finish on all surfaces including chrome, stainless steel and laminate counter tops.
UOM: 1 * 6 Bottle
Catalog Number: (115-3048)
Supplier: DESCO
Description: Reztore® Surface and mat cleaners removes dust, grease, grime, fingerprints, solder flux and other contaminants from ESD mats and other surfaces.
UOM: 1 * 1 items
Supplier: CONTEC
Description: Neutral detergent with purified water. NeutraKlean is a gentle and low-foaming detergent designed for the cleaning of life science cleanrooms.
Catalog Number: (DIVY100894371)
Supplier: Diversey
Description: Detergent suitable for daily cleaning of heavily soiled surfaces, such as ovens and grills.
UOM: 1 * 2 Bottle
Safer Human and Environmental Health
Sustainable Materials
Sustainable Packaging
Waste Reducing
Catalog Number: (DIVY101103248)
Supplier: Diversey
Description: ClearKlens solve is an alkaline foam detergent for use in the pharmaceutical and cosmetic industries.
UOM: 1 * 1 Bottle
Catalog Number: (141-1582)
Supplier: Diversey
Description: Concentrated daily cleaner for all water-resistant hard surfaces, such as toilets, tiles and porcelain in washrooms.
UOM: 1 * 2 Bottle
Safer Human and Environmental Health
Sustainable Materials
Sustainable Packaging
Waste Reducing
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 167
no targeter for Bottom