You Searched For: Standards
5 304  results were found
SearchResultCount:"5304"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: The VWR® range of secondary reference standards offers extraordinary purity and homogeneity, and are perfectly suited for the verification factors for...
Supplier: VWR Chemicals
Description: For spectrophotometer calibration according to European Pharmacopoeia (Ph. Eur.)
Supplier: VWR Chemicals
Description: Cold Filter Plugging Point (CFPP) Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and deriv...
Catalog Number: (85666.180)
Supplier: VWR Chemicals
Description: A secondary GC solvent reference standard for various chromatographic and analytical applications.
UOM: 1 * 100 mL
Catalog Number: (85654.180)
Supplier: VWR Chemicals
Description: A secondary GC solvent reference standard for various chromatographic and analytical applications.
UOM: 1 * 100 mL
Supplier: VWR Chemicals
Description: Avantor now offers a comprehensive range of osmolality standards as Certified Reference Materials (CRM) for the calibration of osmometers.
...
Supplier: VWR Chemicals
Description: Sulphur CRMs are suitable for calibration, verification or for use in quality control procedures used for analysing sulphur content in petroleum produ...
Supplier: VWR Chemicals
Description: Sulphur CRMs are suitable for calibration, verification or for use in quality control procedures used for analysing sulphur content in petroleum produ...
Supplier: VWR Chemicals
Description: Sulphur CRMs are suitable for calibration, verification, or for use in quality control procedures used for analysing sulphur content in petroleum prod...
Supplier: VWR Chemicals
Description: Sulphur CRMs are suitable for calibration, verification, or for use in quality control procedures used for analysing sulphur content in petroleum prod...
Supplier: VWR Chemicals
Description: Total Base Number (TBN) standards are specifically manufactured for the verification of analytical instruments used to determine base number by potent...
Supplier: VWR Chemicals
Description: Sulphur CRMs are suitable for calibration, verification or for use in quality control procedures used for analysing sulphur content in petroleum produ...
Supplier: VWR Chemicals
Description: Total Acid Number (TAN) standards are specifically manufactured for the verification of analytical instruments used to determine acid number by potent...
Supplier: VWR Chemicals
Description: These standards can be used for calibration, control, verification, qualification or method validation of kinematic and dynamic viscosity measurement ...
Supplier: VWR Chemicals
Description: Ready to use colour solutions that are prepared gravimetrically on a weight/weight basis.
Supplier: VWR Chemicals
Description: High quality CCS viscosity standards are ideal for the calibration and verification of analytical equipment used in Cold Cranking Simulator (CCS) test...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 5 304
no targeter for Bottom