You Searched For: Razors and Blades
28  results were found
SearchResultCount:"28"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Made from food grade PET. At just 0,27 mm, the VWR® gel cutter is as thin as a single edge industrial razor blade, but much safer to use.
Catalog Number: (631-1142)
Supplier: VWR Collection
Description: VWR®, Replacement blades for ABS dispenser
UOM: 1 * 5 items
Catalog Number: (HEATHS234525R)
Supplier: HEATHROW
Description: Replacement blades for ABS dispenser
UOM: 1 * 5 items
Supplier: SWANN-MORTON
Description: The KLEEN Blade Management System allows for effortless safe attachment and removal of surgical blades from a standard handle. The system which incorp...
Catalog Number: (RSGA102.052)
Supplier: RSG
Description: Replacement blades, pack of 5
UOM: 1 * 10 items
Catalog Number: (PRTWKN91NCR)
Supplier: Portwest
Description: These blades are constructed from carbon steel.
UOM: 1 * 500 items
Catalog Number: (233-0188)
Supplier: MARTOR ARGENTAX
Description: Concealed replacement blade for Secumax 350.
UOM: 1 * 10 items
Supplier: MARTOR ARGENTAX
Description: Non corrosive blade for mounting in safety knives and cutters.
Catalog Number: (233-2211)
Supplier: MARTOR ARGENTAX
Description: Industrial blade for use with safety knives and cutters.
UOM: 1 * 10 items
Supplier: MARTOR ARGENTAX
Description: Hook blades, 2-sided ground-edge, Steel, Thickness: 0,63 mm, L×W: 51×19 mm
Catalog Number: (233-2238)
Supplier: MARTOR ARGENTAX
Description: Non corrosive trapezoid blade for use with safety knives.
UOM: 1 * 10 items
Catalog Number: (233-0139)
Supplier: MARTOR ARGENTAX
Description: Trapezoid blade for use with safety knives.
UOM: 1 * 10 items
Catalog Number: (MARO86.60)
Supplier: MARTOR ARGENTAX
Description: Styropor blade for use with safety knives.
UOM: 1 * 10 items
Catalog Number: (233-2240)
Supplier: MARTOR ARGENTAX
Description: Industrial blade for use with safety knives.
UOM: 1 * 10 items
Catalog Number: (223-0112)
Supplier: MARTOR ARGENTAX
Description: Trapezoid blade with rounded tips for use with safety knives.
UOM: 1 * 1 SET
Catalog Number: (223-0114)
Supplier: MARTOR ARGENTAX
Description: Non corrosive trapezoid blade for use with safety knives.
UOM: 1 * 10 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 28
no targeter for Bottom