You Searched For: Proteins and Peptides
8 589  results were found
SearchResultCount:"8589"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: SIGMA-ALDRICH MICROSCOPY
Description: Protargol silver impregnation and neuronal selectivity for light and electron microscopic observations.
Supplier: SIGMA-ALDRICH MICROSCOPY
Description: Poly-L-lysine is a positively charged amino acid polymer with approximately one HBr per lysine residue. The hydrobromide allows the poly-L-lysine to b...
Supplier: VWR Chemicals
Description: VWR®, Gelatin
Catalog Number: (422361V)
Supplier: VWR Chemicals
Description: A protease-free grade albumin for RIA and ELISA applications, produced by a heat-shock process in caprylic acid.
UOM: 1 * 100 g
Catalog Number: (421501J)
Supplier: VWR Chemicals
Description: A standard grade albumin for basic applications, produced by a heat shock process in caprylic acid.
UOM: 1 * 100 g
Catalog Number: (422351S)
Supplier: VWR Chemicals
Description: A higher purity product than standard grade or Cohn fraction V material. It is negative for globulins that interfere in many assays.
UOM: 1 * 100 g
Supplier: VWR Chemicals
Description: Fluorescence: 238 to 282
E. coli: equivalent
Water content: < or > 13%
pH at 25 °C: 4,0 - 5,7
Salmonella: not detec...
Catalog Number: (M183-5MG)
Supplier: VWR Chemicals
Description: An aspartic protease inhibitor of pepsin, renin, and cathepsin D. Can also block HIV protease. Not soluble in water.
UOM: 1 * 5 mg
MSDS
Supplier: VWR Chemicals
Description: A reversible inhibitor of cysteine proteases and serine proteases resembling trypsin. Inhibits trypsin, plasmin, papain, kallikrein, thrombin, and cat...
Catalog Number: (422371X)
Supplier: VWR Chemicals
Description: A Cohn fraction V albumin produced by cold ethanol fractionation with minimum use of chemicals. Fatty acids and native sialic acid residues are preser...
UOM: 1 * 100 g
Supplier: VWR Chemicals
Description: A 60 kDa tetrameric protein that exhibits high binding affinity for biotin. Able to bind one molecule of biotin with each subunit, streptavidin is use...
Catalog Number: (0440-01)
Supplier: Avantor
Description: J.T.Baker®, Chicken Native Albumin (from Egg)
UOM: 1 * 500 g
Catalog Number: (E429-50MG)
Supplier: VWR Chemicals
Description: A competitive and reversible serine protease inhibitor of chymotrypsin, trypsin, kallikrein and plasmin. Not an inhibitor of Factor Xa and thrombin.
UOM: 1 * 50 mg
Catalog Number: (422381B)
Supplier: VWR Chemicals
Description: Produced by a heat shock process using sodium caprylate as a stabiliser with additional treatments including dialysis and deionisation.
UOM: 1 * 100 g
Supplier: VWR Chemicals
Description: High molecular weight phosphoprotein prepared from milk.
Catalog Number: (22544.292)
Supplier: VWR Chemicals
Description: Proteins (on anhydrous product): Min. 92,5 %
Free acidity: Max. 1,0%
Fats: Max. 2,5%
Ignition residue (SOâ‚„): Max. 2,75%
Water: Max...
UOM: 1 * 1 kg
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 8 589
no targeter for Bottom