You Searched For: Petroleum Testing Apparatus
4  results were found
SearchResultCount:"4"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (SCERSP513880-0000)
Supplier: DWK Life Sciences
Description: The Filter Stick Apparatus is used in the determination of oil content and solvent extractables in petroleum waxes.
UOM: 1 * 1 items
Catalog Number: (201-1419)
Supplier: DWK Life Sciences
Description: Ramsbottom coking bulbs are used in the determination of carbon residue in petroleum products.
UOM: 1 * 25 items
Catalog Number: (HOFM40052.00)
Supplier: HOFMANN GLASTECHNIK GMBH
Description: Apparatus for the determination of essential oil.
UOM: 1 * 1 items
Catalog Number: (NEUB2749-90-010)
Supplier: NEUBERT VOLUM. GLASSWAERE
Description: This Ross-Miles apparatus is used for testing of surfactants.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 4 of 4
no targeter for Bottom