Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "paraffin"
https://si.vwr.com/stibo/smallweb/std.lang.all/96/31/18989631.jpg
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"4"
  List View Grid View Easy View (new)
Sort by:
Catalog Number: (10047400.)
Supplier:  VWR Chemicals
Description:   Solvent replacement for cleaning instruments and equipments in contact with paraffin wax. paraffin wax is used to prepare histological sections for mi...
UOM:  1 * 100 ml
Catalog Number: (301440ZK)
Supplier:  VWR Chemicals
Description:   Sterile liquid paraffin is mostly used in microbiology as a vehicle to improve the anaerobic culture media as : O / F, decarboxylase medium, MIO, etc....
UOM:  1 * 100 ml
Catalog Number: (VWRK2079-A)
Supplier:  KLINIPATH (PL)
Description:   Highly purified histology wax for processing and embedding.
UOM:  1 * 12 kg
Supplier:  LEICA MICROSYSTEMS
Description:   Paraplast® is recommended for general tissue embedding. It is composed of a refined mixture of highly purified paraffin containing plastic polymers of...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 4  of 4