Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Mirrors
https://si.vwr.com/stibo/smallweb/std.lang.all/95/54/18989554.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"7"
  List View Grid View Easy View (new)
Sort by:
Catalog Number: (SIMELD3025)
Supplier:  TQC
Description:   Ideal for visual inspection of hard-to-reach spots. Spots that are difficult to inspect are even more difficult to blast or coat.
UOM:  1 * 1 items
Catalog Number: (SIMELD3004)
Supplier:  TQC
Description:   Perfect tool to use for inspection of professional coating jobs on large structures, ships, tanks, buildings etc.. The inspection mirrors offers a cle...
UOM:  1 * 1 items
Catalog Number: (MICRQDIST-66P)
Supplier:  MICRONOVA
Description:   Inspection Mirror has a swivel connection allowing it to be attached to stainless steel handle and allows the operator to run the unit behind equipmen...
UOM:  1 * 1 items
Supplier:  DIDALAB
Description:   Mirrors used for enabling classroom experiments.
Supplier:  Tinman
Description:   These wall mounted cleanroom mirrors complement the gowning area of the cleanroom.
Supplier:  AESCULAP AG & CO
Description:   MIRROR LARYNGEAL SIZE 7.24MM DIAM. 1 * 1 items
Supplier:  AESCULAP AG & CO
Description:   The mouth mirror casts fine image without reflections under mid luminous conditions in the case of standard mirrors and high luminous conditions in th...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 7  of 7
A new look and a renewed commitment
Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.
Learn More Start Shopping