You Searched For: Gas Regulators
17  results were found
SearchResultCount:"17"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (HYPE60180-926)
Supplier: Thermo Fisher Scientific
Description: High purity brass gas regulators are used for non-corrosive analytical and process applications.
UOM: 1 * 1 items
Catalog Number: (REST21666)
Supplier: Restek
Description: Chromium plated brass line regulator.
UOM: 1 * 1 items
Catalog Number: (REST22368)
Supplier: Restek
Description: This gas regulator is ultra high purity chrome plated brass coated with international fittings for sensitive GC applications using MS, PID, or ECD det...
UOM: 1 * 1 items
Supplier: Agilent
Description: These gas regulators ensure that ICP-MS applications and methods are carried out with the consistent and stable gas flows. Effective gas regulation is...
Supplier: Restek
Description: These air gas regulators are made of brass body, Viton® seal and diaphragm, acetal seat.
Supplier: Restek
Description: Singlel stage gas regulator, for Air
Supplier: Restek
Description: Ultra high purity gas regulators are ideal for sensitive GC applications using MS, PID and ECD detection methods. They feature reduced internal dead v...
Supplier: Restek
Description: This compact general purpose regulator is constructed of lightweight aluminum with an elastomer diaphragm. It has many laboratory applications includi...
Supplier: Restek
Description: Automatic switchover system with line regulator, CGA 350 (Hâ‚‚, Pâ‚…) fitting
Supplier: Restek
Description: These regulators are the standard for ultra high purity and corrosion resistant pressure regulation. They are more easily purged of atmospheric compon...
Supplier: Restek
Description: These single stage stainless steel VOC regulators features stainless steel diaphragm, two pressure gauges, CGA-180 fitting and Kel-F® seat.
Supplier: Restek
Description: These single stage stainless steel VOC regulators feature stainless steel diaphragm, two pressure gauges, CGA-180 fitting and Kel-F® seat.
Catalog Number: (REST20634)
Supplier: Restek
Description: This backpressure regulator kit increases the versatility of Agilent 5890 GC by replacing the existing 0,06 bar (207 kPa) backpressure regulator and g...
UOM: 1 * 1 items
Catalog Number: (REST22032)
Supplier: Restek
Description: This mini regulator is suitable for natural gas and refinery gas standards.
UOM: 1 * 1 items
Catalog Number: (REST21334)
Supplier: Restek
Description: This hydrogen flashback arrestor is a special gas safety device, it cuts to stop the flame or reverse flow of gas back up into the equipment or supply...
UOM: 1 * 1 items
Supplier: Restek
Description: Singlel stage gas regulator, for N2, He, Ar
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 17
no targeter for Bottom