You Searched For: Deodorants and Air Fresheners
11  results were found
SearchResultCount:"11"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Chemicals
Description: This autoclave deodorant improves the working environment and neutralises odours associated with autoclaving.
Catalog Number: (BELAH131990000)
Supplier: Bel-Art Products, a Part of SP
Description: This heat-released deodorant pads neutralise autoclave odors with a pleasant scent.
UOM: 1 * 100 items
Catalog Number: (115-2059)
Supplier: KIMBERLY CLARK
Description: The air care fragrances create a clean, fresh and constantly pleasant washroom environment. Dispensers need to be ordered separately.
UOM: 1 * 6 items
Supplier: Essity
Description: A1 aerosol dispenser, plastic, grey, 84×66×168 mm, Tork®
Supplier: Essity
Description: Aerosol refill for A1 air freshener, floral fragrance, Tork® Premium
Supplier: Bel-Art Products, a Part of SP
Description: These pads are impregnated with a deodorant that is released by heat, which doesn't merely mask autoclave odours but actually neutralises them.
Supplier: Essity
Description: Air freshening system consisting of an impregnated rubber tag that delivers a fresh, clean fragrance.
Catalog Number: (KING070518)
Supplier: KING DISPOSABLES
Description: This room fragrance neutralises bad odours and diffuses a pleasant lavender fragrance throughout the entire room.
UOM: 1 * 1 items
Catalog Number: (115-1699)
Supplier: Essity
Description: Tork Air Fresheners eliminate unpleasant odours in washrooms. The starter pack contains a dispenser (incl. batteries) and the three aerosols Floral, C...
UOM: 1 * 1 KIT
Catalog Number: (KING070540)
Supplier: KING DISPOSABLES
Description: Odor neutralising spray for toilets, locker rooms, hospitals and more.
UOM: 1 * 12 items
Catalog Number: (115-1688)
Supplier: Essity
Description: Dispenser / Holder for A2 air freshener disc, Tork Universal, 75×64×120 mm, Tork®
UOM: 1 * 4 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 11 of 11
no targeter for Bottom