Vitality hrGFP mammalian expression
Supplier: Agilent Technologies
AGLS240059EA 774 EUR
AGLS240059 AGLS240043
Vitality hrGFP mammalian expression
Vitality hrGFP (humanised, Renilla reniformis green fluorescent protein) products are optimal for mammalian expression protein studies that examine gene expression, transfection efficiency, and protein localisation.
  • Applications include Western blotting, immunoprecipitation and flow cytometry
This hrGFP is derived from a different organism than EGFP, so the hrGFP antibody does not cross react with the Aequorea victoria GFP variant-derived EGFP.
Upozornení: For Research Use Only. Not for Use in Diagnostic Procedures.