Order Entry
Quoirin and Lepoivre basal salt mixture
Quoirin and Lepoivre basal salt mixture
Supplier:  Apollo Scientific
Certificates
Quoirin and Lepoivre basal salt mixture
Supplier:  Apollo Scientific
Specifications
  • Environmentally Preferable:
Specifications
Product Information
  • Catalog #
    Supplier #
    Seq
    Pk
    Availability
    Price
  • Quoirin and Lepoivre basal salt mixture 10×1 L
    Quoirin and Lepoivre basal salt mixture 10×1 L
    APOSPMQ673A-10X1L
    PMQ673A-10X1L
    01
    10
    Log in for availability
    1 * 10 items
    1 * 10 items - 26,40 €
    26,40 €
    New Customer Promotional Price
    1 * 10 items
       26,40 €
  • Quoirin and Lepoivre basal salt mixture 10 L
    Quoirin and Lepoivre basal salt mixture 10 L
    APOSPMQ673B-10L
    PMQ673B-10L
    02
    10 L
    Log in for availability
    1 * 10 L
    1 * 10 L - 21,90 €
    21,90 €
    New Customer Promotional Price
    1 * 10 L
       21,90 €
  • Quoirin and Lepoivre basal salt mixture 50 L
    Quoirin and Lepoivre basal salt mixture 50 L
    APOSPMQ673C-50L
    PMQ673C-50L
    03
    50 L
    Log in for availability
    1 * 50 L
    1 * 50 L