Anti-FLAG® Anti-DYKDDDDK Antibody
Supplier: Agilent Technologies
AGLS200474EA 411 EUR
AGLS200474 AGLS200473
Anti-FLAG® Anti-DYKDDDDK Antibody
Antibodies
The FLAG® peptide (DYKDDDDK) is widely used as an epitope tag for protein expression analysis, purification, and detection.
  • Rat monoclonal antibody to DYKDDDDK epitope tag
  • High specificity – reduced background and less cross-reactivity
  • Highly sensitivity – strong signal and ability to detect low amounts of tag
The Anti-FLAG® M2 Antibody is optimized for the detection of FLAG-fusion proteins in mammalian, plant and bacterial expression systems. This antibody provides equivalent sensitivity and specificity to similar antibodies.
Upozornení: For research use only. Not for use in diagnostic procedures.