Aquabator-Clean™ (100X)
Supplier: Apollo
APOSBIA9390-250MLEA 63 EUR
APOSBIA9390-250ML
Aquabator-Clean™ (100X)
Cleaning Agents Water Bath Protection Reagents
Disinfectant solution for ordinary water baths (not for CO2-incubators)
  • Prevention of microbial growth in water baths
  • 100X concentrated solution (Use 10 ml per 1 liter of water.)
Upozornení: Use biocides safely. Always read the label and product information before use.
Aquabator-Clean™ is a registered trademark of AppliChem GmbH.