Disinfectant solution Incubator-Clean™
Supplier: Apollo
APOSBIA5230-500MLEA 0 EUR
APOSBIA5230-500ML APOSBIA5230-5L
Disinfectant solution Incubator-Clean™
Cleaning Agents Disinfectants
Incubator-Clean™ disinfectant solution prevents contamination with, and growth of, fungi (and spores), bacteria (and spores, including Tuberculosis bacteria), viruses (including HIV and Hepatitis B) and mycoplasma in incubators and/or on sterile work benches. The active ingredients are quaternary benzylammonium compounds.
Upozornení: Use biocides safely. Always read the label and product information before use.
Incubator-Clean™ is a registered trademark of AppliChem GmbH.