Incuwater-Clean™
Supplier: Apollo
APOSBIA5219-100MLEA 68.4 EUR
APOSBIA5219-100ML
Incuwater-Clean™
Cleaning Agents Water Bath Protection Reagents
Disinfectant solution for CO2-incubator water baths, for the prevention of microbial growth in incubator water baths.
  • 100X concentrated solution.
Use 50 ml per 5 liter of incubator water bath.
Upozornení: Incuwater-Clean™ is a registered trademark of AppliChem GmbH.