3-Ethynylpyridazine
Supplier: Apollo Scientific
OR942624-1G
OR942624-250MG
APOSOR942624-1GEA
487
EUR
APOSOR942624-1G
APOSOR942624-250MG
3-Ethynylpyridazine
3-Ethynylpyridazine
Formula:
C₆H₄N₂ MW: 104.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16988473 CAS Number: 1017793-08-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...