2-Amino-3-methylpyridine 97%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 3-Methyl-2-pyridylamine, 3-Methylpyridin-2-amine, 2-Amino-3-picoline
CAS Number:1603-40-3
:
Formula:C₆H₈N₂
MDL Number:MFCD00006320
Boiling Pt:221…222 °C
Melting Pt:29…32 °C
Density:1.073 g/cm³ (25 °C)
Molecular Weight:108.14 g/mol
Flash Pt:118 °C
Storage Temperature:Ambient
EINECS:216-501-4
UN:2811
ADR:6.1 III
:GHS06
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Product Information
- Catalog #Supplier #SeqPack typePkAvailabilityPrice