2-Hydroxyacetamide

Supplier: Apollo Scientific
Warning

OR21888-25G
APOSOR21888-25GEA 115 EUR
APOSOR21888-25G
2-Hydroxyacetamide
2-Hydroxyacetamide
Formula: C₂H₅NO₂
MW: 75.07 g/mol
Melting Pt: 118…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00047895
CAS Number: 598-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR