Cleanroom gloves, nitrile, CERTICLEAN®
Supplier: KIMBERLY CLARK
Certificates
About this item
Ambidextrous gloves with beaded cuffs made from silicone-free nitrile. The specialised manufacturing processes result in consistently low particle counts and low levels of ionic extractables and non-volatile residue (NVR) transfer.
- Non sterile, powder-free, textured fingertips
- In-use static dissipative properties are superior to natural rubber latex
- Tapered cuff provides more coverage over the garment sleeve
Suitable for use in Class 10, ISO 4, M2.5 and lower cleanrooms.
Packaging: Packaged in cleanroom-compatible polyethylene, 100 gloves/double poly bag, 10 bags/case.