You Searched For: Sample Containers
156  results were found
SearchResultCount:"156"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Q Path® containers (PP) with suitable caps (PE) for safe transport of biopsy probes.
Supplier: VWR Collection
Description: PP, clear, with LDPE screw cap.
Supplier: VWR Collection
Description: PP.
Supplier: VWR Collection
Description: PP, translucent.
Supplier: VWR Collection
Description: PP, translucent.
Catalog Number: (720-1987)
Supplier: VWR Collection
Description: Container for transport of biological samples.
UOM: 1 * 120 items
Supplier: VWR Collection
Description: PP, translucent.
Supplier: KARTELL
Description: PP, translucent or PS, transparent with PE lid.
Supplier: DWK Life Sciences
Description: Polystyrene clear, with metal screw cap.
Supplier: DWK Life Sciences
Description: PS clear, with PE screw cap.
Supplier: KARTELL
Description: Made of PS with PE screw cap and spoon.
Supplier: BOCHEM
Description: Made of 18/10 stainless steel.
Supplier: KINEMATICA
Description: Dispersing vessel, cloverleaf-like, without cap, Borosilicate glass, Capacity: 1000 ml, Ø ext.: 90 mm, Height: 250 mm, GS 50
Supplier: KARTELL
Description: PS, transparent.
Supplier: Brand
Description: PS, transparent.
Supplier: Kautex
Description: PVC, square, transparent or brown transparent colours.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 156
no targeter for Bottom