VWR®, Speciality Filter Tips
Supplier: Avantor
613-0996EA 118 EUR
VWRI613-0996
VWR®, Speciality Filter Tips
PP, sterile.
  • Sterilised by gamma irradiation
  • Certified RNase-, DNase-, DNA- and pyrogen-free