You Searched For: Pens
49  results were found
SearchResultCount:"49"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: These versatile laboratory markers are designed to write on wet or dry surfaces, and are suitable for industrial, biomedical and educational laborator...
Supplier: VWR Collection
Description: Perfect for slides, sample bags and glassware.
Catalog Number: (631-1136)
Supplier: VWR Collection
Description: The ultra-fine tip of this permanent marker, and the contrasting black colour, allow clearly legible writing. Writes on glass, plastic and paper.
UOM: 1 * 12 items
Catalog Number: (HECH41182010)
Supplier: GLASWARENFABRIK KARL HECHT
Description: Diamond pen, Application: Glass
UOM: 1 * 1 items
Supplier: KEYSTONE CLEANROOM PRODUCTS
Description: These pens and markers eliminate the risks associated with surface sanitisation procedures.
Sale
Supplier: KEYSTONE CLEANROOM PRODUCTS
Description: These pens and markers eliminate the risks associated with surface sanitisation procedures.
Sale
Supplier: Thermo Fisher Scientific
Description: For labelling cryomaterials such as cryotubes, cryoboxes and cryolabels. Set contains one of each colour pen: black, blue, red and green.
Catalog Number: (BRDY335091)
Supplier: Brady
Description: These labelling pens use a quick drying ink.
UOM: 1 * 5 items
Catalog Number: (SIMPM795-1)
Supplier: Simport Scientific
Description: The microscope diamond slide writer is compatible with a wide variety of slide labellers.
UOM: 1 * 6 items
Supplier: OUTILS RUBIS SA
Description: Hexagonal aluminium scribing pen, 8 mm, with replaceable diamond tip.
Catalog Number: (BRDY110594)
Supplier: Brady
Description: These marker pens are erasable.
UOM: 1 * 4 items
Supplier: MICRONOVA
Description: A permanent marker that gives the thinnest permanent line at 0,3 mm.
Supplier: MICRONOVA
Description: Micronova cleanroom pens feature low sodium, indelible ink and a special low particulate barrel to limit particle generation. Cleanroom irradiated Sha...
Supplier: MICRONOVA
Description: LabMarkers feature permanent ink that marks on cold or wet surfaces, plastic bags or disposable labware. Marks will not smear on reusable glass, metal...
Supplier: MICRONOVA
Description: The industry standard, original pen style, fine point, permanent marker.
Catalog Number: (811-0054)
Supplier: EPPENDORF
Description: This pen is ideal for inscribing on microtest tubes, centrifuge tubes and more.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 49
no targeter for Bottom